Learn More
Biomatik Corporation Human Epidermal Growth Factor Receptor, Egfr Elisa Kit
Supplier:  Biomatik Corporation EKC31215
Encompass_Preferred
Human Epidermal Growth Factor Receptor, Egfr Elisa Kit for detecting Epidermal Growth Factor Receptor (Erythroblastic Leukemia Viral (V-Erb-B) Oncogene Homolog, Avian) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.019ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00533; EGFR. Target Synonyms: Avian erythroblastic leukemia viral (v erb b) oncogene homolog; Cell growth inhibiting protein 40; Cell proliferation inducing protein 61; EGF R; EGFR; EGFR_HUMAN; Epidermal growth factor receptor (avian erythroblastic leukemia viral (v erb b) oncogene homolog); Epidermal growth factor receptor (erythroblastic leukemia viral (v erb b) oncogene homolog avian); Epidermal growth factor receptor; erb-b2 receptor tyrosine kinase 1; ERBB; ERBB1; Errp; HER1; mENA; NISBD2; Oncogen ERBB; PIG61. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7418