Learn More
Biomatik Corporation Human Epidermal Growth Factor (EGF) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12396
Encompass_Preferred
Human Epidermal Growth Factor (EGF) High-sensitive ELISA Kit for detecting EGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01133. Gene ID: 1950. Target Synonyms: URG; Beta-Urogastrone; Pro-epidermal growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7971