Learn More
Biomatik Corporation Human Epigen (EPG) CLIA Kit
Supplier:  Biomatik Corporation EKU09734
Encompass_Preferred
Human Epigen (EPG) CLIA Kit for detecting EPG in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q6UW88. Gene ID: 255324. Target Synonyms: EPGN; Epithelial Mitogen Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7350