Learn More
American Research Products Inc Human Epithelial neutrophil ac
Catalog No. 5000326515
Encompass
Human Epithelial neutrophil actiVating peptide 78 (ENA-78/CXCL5) ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 567 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26515 Supplier American Research Products Inc Supplier No. CSBE08178H
Human Epithelial neutrophil actiVating peptide 78 (ENA-78/CXCL5) ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 567 ng/mL Sample serum plasma tissue homogenates Method Sandwich