Learn More
Sino Biological Human EPO Protein (Fc Tag)
Supplier:  Sino Biological 10236H02H1
Encompass_Preferred
A DNA sequence encoding the human EPO (NP_000790.2) (Met1-Arg193) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5415