Learn More
Sino Biological Human EPOR MAb 20UL 10707-R016-20
Supplier:  Sino Biological 10707R01620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EPOR / Erythropoietin Receptor (rh EPOR / Erythropoietin Receptor; Catalog#10707-H08H; NP_000112.1; Met1-Pro250).
Catalog No. 50-240-8412