Learn More
Sino Biological Human EPOR MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10707MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EPOR/Erythropoietin Receptor (rh EPOR/Erythropoietin Receptor; Catalog#10707-H08H; NP_000112.1; Met1-Pro250). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1298