Learn More
Sino Biological Recombinant Human Erythropoietin R/EPOR Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10707H08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Ala25-Pro250) of human EPOR (NP_000112.1) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5194