Learn More
Bioworld HUMAN EPTIFIBATIDE
Supplier:  Bioworld 220603101
Encompass_Preferred
Eptifibatide Description : Synthetic Eptifibatide is a 6 amino acid peptide having a MW of 832 dalton the following sequence Mpr-Har-Gly-Asp-Trp-Pro-Cys-NH2 .Eptifibatide binds to the platelet receptor glycoprotein(GP) IIb/IIIa of huma...
Catalog No. 50-253-408