Learn More
Sino Biological Human ERAP2 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10734H07H50
Encompass_Preferred
A DNA sequence encoding the lumenal domain of human ERAP2 (NP_071745.1) (Ala 56-Thr 960) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3116