Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (Site-Specific PE Conjugation, ECD, His Tag)
Catalog No. 502993134
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Thr652) of human ERBB2 (NP_004439.2) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with PE (Excitation = 495-566 nm, Emission Max.= 575 nm).
Catalog No. 50-299-3134 Supplier Sino Biological Supplier No. 10004H86HSP25
A DNA sequence encoding the extracellular domain (Met1-Thr652) of human ERBB2 (NP_004439.2) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with PE (Excitation = 495-566 nm, Emission Max.= 575 nm).