Learn More
Bon Opus Biosciences Recombinant Human ERBB2 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP109850UG
Encompass_Preferred
Recombinant Human ERBB2 (C-His) is produced by E.coli. The target gene encoding Cys511-Pro644 is expressed with a C-His tag. Accession# P04626. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1098
Catalog No. 50-217-2696