Learn More
Sino Biological Recombinant Anti-Her2/ERBB2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10004R002A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human HER2/ErbB2/CD340 (rh HER2/ErbB2/CD340; Catalog#10004-H08H; NP_004439.2; Met1-Thr652).
Catalog No. 50-241-1570