Learn More
Sino Biological Anti-Her2/ERBB2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10004MM42100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Her2/ERBB2 (rh Her2/ERBB2; Catalog#10004-H20B1; NP_004439.2; Lys676-Val1255). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9005