Learn More
Sino Biological Human ERBB2 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10004R51120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ErbB2 / HER2 extracellular domain (rh ErbB2; Catalog#10004-H08H; NP_004439.2; Met 1-Thr 652).
Catalog No. 50-237-1689