Learn More
Sino Biological Anti-Her2/ERBB2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 206304T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Her2 / ERBB2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-8342