Learn More
Sino Biological Anti-Her2/ERBB2 Antibody (Biotin), Rabbit Polyclonal
Supplier:  Sino Biological 10004RP03B20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ErbB2 / HER2 extracellular domain (rhErbB2; Catalog#10004-H08H; NP_004439.2; Met 1-Thr652). Total IgG was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-255-5540