Learn More
Sino Biological Human ERBB2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10004AT54100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant extracellular domain Human Her2/ERBB2 (rh Her2/ERBB2; Catalog#10004-H08H; NP_004439.2; Met1-Thr652). Her2/ERBB2 specific IgG was purified by Human Her2/ERBB2 affinity chromatography.
Catalog No. 50-237-3186