Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10004H02HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Thr652) of human ErbB2 (NP_004439.2) was fused with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5549