Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10004H08H4B20
Encompass_Preferred
A DNA sequence encoding the human ERBB2 (AAA75493.1) (Pro489-Cys630) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5545