Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (ECD, domain IV, His Tag)
Supplier:  Sino Biological 10004H08H4500
Encompass_Preferred
A DNA sequence encoding the human ERBB2 (NP_004439.2) (Pro489-Cys630) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5546