Learn More
ACROBiosystems ACROBiosystems Human ErbB3 / Her3 Protein, Fc Tag
Catalog No. 502106634
Encompass_Preferred
Recombinant Protein;1MG;Human ErbB3, Fc Tag (ER3-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 94.8 kDa. The protein migrates as 116-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ErbB3, Fc Tag (Cat. No. ER3-H5259) at 2 μg/mL (100 μL/well) can bind Human NRG1-beta 1 Protein with a linear range of 1-20 ng/mL (Routinely tested).
Catalog No. 50-210-6634 Supplier ACROBiosystems Supplier No. ER3H52591MG
Recombinant Protein;1MG;Human ErbB3, Fc Tag (ER3-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 94.8 kDa. The protein migrates as 116-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ErbB3, Fc Tag (Cat. No. ER3-H5259) at 2 μg/mL (100 μL/well) can bind Human NRG1-beta 1 Protein with a linear range of 1-20 ng/mL (Routinely tested).