Learn More
Sino Biological Recombinant Human HER3/ERBB3 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10201H02HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Thr643) of human ErbB3 (NP_001973.2) precursor was expressed with the C-terminal fused Fc region of human IgG1. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5441