Learn More
Sino Biological Recombinant Human HER3/ERBB3 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10201H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Thr643) of human ErbB3 (NP_001973.2) precursor was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5438