Learn More
Sino Biological Human ERBB3 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 13346RP01100
Encompass_Preferred
A DNA sequence encoding the Human ERBB3 (NP_001973.2) (Met1-Thr643) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-4281