Learn More
BIOHIPPO Human ERFE Microsample
SDP
Catalog No. 5000313723
Encompass
Human ERFE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ERFE in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ERFE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ERFE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ERFE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13723 Supplier BIOHIPPO Supplier No. BHE15210782
Human ERFE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ERFE in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ERFE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ERFE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ERFE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color