Learn More
Sino Biological Human ERH PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14483T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ERH (rh ERH; Catalog#14483-H07E; P84090; Met1-Lys104). ERH specific IgG was purified by Human ERH affinity chromatography.
Catalog No. 50-237-1505