Learn More
Sino Biological Human ERN1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11905T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IRE1 (rh IRE1; Catalog#11905-H20B; O75460-1; Pro465-Leu977). IRE1 specific IgG was purified by Human IRE1 affinity chromatography.
Catalog No. 50-237-0880