Learn More
Biomatik Corporation Human Erythrocyte Membrane Protein Band 4.2 (EPB42) ELISA Kit
Supplier:  Biomatik Corporation EKU03959
Encompass_Preferred
Human Erythrocyte Membrane Protein Band 4.2 (EPB42) ELISA Kit for detecting EPB42 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16452. Gene ID: 2038. Target Synonyms: PA; Erythrocyte Surface Protein Band 4.2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6505