Learn More
Abcam Human Erythropoietin ELISA Kit (EPO), 10 x 96 Tests
Supplier:  Abcam AB27439710X96TESTS
Encompass_Preferred
Human Erythropoietin ELISA Kit (EPO) is a single-wash 90-min SimpleStep Sandwich ELISA used to quantify Human Erythropoietin (EPO) with a sensitivity of 1 mIU/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout, works on any plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well for higher throughput - Need to design your own assay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-5922
Only null left
Add to Cart
Request a Quote