Learn More
ACROBiosystems Human Erythropoietin R 100ug
Catalog No. 502535089
Encompass_Preferred
Human Erythropoietin R Protein Fc Tag (ERR-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Pro 250 (Accession # P19235-1).
Catalog No. 50-253-5089 Supplier ACROBiosystems Supplier No. ERRH5253100UG
Human Erythropoietin R Protein Fc Tag (ERR-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Pro 250 (Accession # P19235-1).