Learn More
Sino Biological Human Erythropoietin R/EPOR Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785607
Encompass_Preferred
The Human EPOR ELISA Pair Set is for the quantitative determination of Human EPOR.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for EPOR coated on a 96-well plate. Standards and samples are added to the wells, and any EPOR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-EPOR monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of EPOR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85607 Supplier Sino Biological Supplier No. SEK107075
The Human EPOR ELISA Pair Set is for the quantitative determination of Human EPOR.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for EPOR coated on a 96-well plate. Standards and samples are added to the wells, and any EPOR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-EPOR monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of EPOR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.