Learn More
Biomatik Corporation Human E-selectin (E-selectin) CLIA Kit
Supplier:  Biomatik Corporation EKU08775
Encompass_Preferred
Human E-selectin (E-selectin) CLIA Kit for detecting E-selectin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.63pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P16581. Gene ID: 6401. Target Synonyms: CD62E; CD62-E; E-Selectin; ELAM; ELAM1; ESEL; SEL-E; E-LAM; E-LAM1; E-SEL; LECAM2; Endothelial Leukocyte Adhesion Molecule 1; CD62 Antigen-Like Family Member E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6754