Learn More
Cusabio Technology LLC Human EstradiolE2 ELISA Kit
Catalog No. NC3654213
Encompass_Preferred
This Human Estradiol (E2) ELISA Kit was designed for the quantitative measurement of Human Estradiol (E2) protein in serum, plasma. It is a Competitive ELISA kit, its detection range is 40 pg/mL-1000 pg/mL and the sensitivity is 40 pg/mL.
Catalog No. NC3654213 Supplier Cusabio Technology LLC Supplier No. CSBE05108H
This Human Estradiol (E2) ELISA Kit was designed for the quantitative measurement of Human Estradiol (E2) protein in serum, plasma. It is a Competitive ELISA kit, its detection range is 40 pg/mL-1000 pg/mL and the sensitivity is 40 pg/mL.