Learn More
American Research Products Inc Human Estradiol receptor ER E
Catalog No. 5000329589
Encompass
Human Estradiol receptor ER ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29589 Supplier American Research Products Inc Supplier No. CSBE08652H
Add to Cart
Add to Cart
Human Estradiol receptor ER ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich