Learn More
Biomatik Corporation Human Estrogen Receptor Beta (ERb) CLIA Kit
Supplier:  Biomatik Corporation EKU09838
Encompass_Preferred
Human Estrogen Receptor Beta (ERb) CLIA Kit for detecting ERb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q92731. Gene ID: 2100. Target Synonyms: ER-B; ESRB; ESTRB; NR3A2; NR3-A2; ESR2; Estrogen Receptor 2; Nuclear Receptor Subfamily 3,Group A,Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6062