Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Estrogen Receptor beta (aa 55-70) Synthetic Peptide

Catalog No. PEP011 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP011 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP011 Supplier Invitrogen™ Supplier No. PEP011
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

PEP-011 is a 16 amino acid (amino acids 55-70) synthetic peptide whose sequence is from the N-terminal end of the human estrogen receptor b (ERb). The sequence of this peptide is (amino to carboxy terminus): A-E-P-Q-K-S-P-W-C-E-A-R-S-L-E-H This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and the ERb, catalog # PA1-311. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 mL of distilled water.

Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms dominantly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2100
Species Human
Purification Method purified
Accession Number Q92731
Common Name Estrogen Receptor beta
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation Peptide with no preservative
For Use With (Application) Control
Gene Alias ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor
Gene Symbol ESR2
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Sequence A - E - P - Q - K - S - P - W - C - E - A - R - S - L - E - H
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.