Learn More
Biomatik Corporation Human Eukaryotic Translation Initiation Factor 2 Alpha Kinase 4 (EIF2aK4) ELISA Kit
Supplier:  Biomatik Corporation EKU12543
Encompass_Preferred
Human Eukaryotic Translation Initiation Factor 2 Alpha Kinase 4 (EIF2aK4) ELISA Kit for detecting EIF2aK4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P2K8. Gene ID: 440275. Target Synonyms: GCN2; GCN2-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4935