Learn More
BIOHIPPO Human EVPL Microsample
SDP
Catalog No. 5000313865
Encompass
Human EVPL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EVPL in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human EVPL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human EVPL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human EVPL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13865 Supplier BIOHIPPO Supplier No. BHE15211890
Human EVPL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of EVPL in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human EVPL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human EVPL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human EVPL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color