Learn More
Biomatik Corporation Human Excitatory Amino Acid Transporter 3 (EAAT3) ELISA Kit
Supplier:  Biomatik Corporation EKU03994
Encompass_Preferred
Human Excitatory Amino Acid Transporter 3 (EAAT3) ELISA Kit for detecting EAAT3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43005. Gene ID: 6505. Target Synonyms: SLC1A1; EAAC1; Solute Carrier 1 Member 1; Sodium-dependent glutamate/aspartate transporter 3; Excitatory amino-acid carrier 1; Neuronal and epithelial glutamate transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8991