Learn More
Biomatik Corporation Human Exosome Component 2 (EXOSC2) ELISA Kit
Supplier:  Biomatik Corporation EKU03997
Encompass_Preferred
Human Exosome Component 2 (EXOSC2) ELISA Kit for detecting EXOSC2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13868. Gene ID: 23404. Target Synonyms: RRP4; Rrp4p; hRrp4p; p7; Ribosomal RNA-processing protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7627