Learn More
Biomatik Corporation Human Extracellular Signal Regulated Kinase 2 (ERK2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12349
Encompass_Preferred
Human Extracellular Signal Regulated Kinase 2 (ERK2) Mini sample ELISA Kit for detecting ERK2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.88-3,000pg/ml. Sensitivity: 17.15pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28482. Gene ID: 5594. Target Synonyms: MAPK1; P38; P40; ERK2; ERT1; MAPK2; P42MAPK; P42-MAPK; PRKM1; PRKM2; P41; P41mapk; P41-mapk; Mitogen-Activated Protein Kinase 1; Mitogen-activated protein kinase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4679