Learn More
American Research Products Inc Human EXtracellular superoXide
Catalog No. 5000328066
Encompass
Human EXtracellular superoXide dismutase [Cu-Zn] (SOD3) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates saliva urine Method Sandwich
Catalog No. 50-003-28066 Supplier American Research Products Inc Supplier No. CSBEL022399HU
Human EXtracellular superoXide dismutase [Cu-Zn] (SOD3) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates saliva urine Method Sandwich