Learn More
Sino Biological Human EZR PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12858T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Ezrin / villin 2 (rh Ezrin / villin 2; Catalog#12858-H07E; P15311; Pro2-Leu586). Ezrin / villin 2 specific IgG was purified by Human Ezrin / villin 2 affinity chromatography.
Catalog No. 50-237-2997