Learn More
Sino Biological Recombinant Anti-Factor X Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11076R05820
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Coagulation Factor X/F10 (rh Coagulation Factor X/F10; Catalog#11076-H08B; NP_000495.1; Met1-Lys488).
Catalog No. 50-240-8726