Learn More
Sino Biological Anti-Factor X Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11076RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Coagulation Factor X / FX (rh FX; Catalog#11076-H08B; NP_000495.1; Met 1-Lys 488). Coagulation Factor X / FX specific IgG was purified by Human Coagulation Factor X / FX affinity chromatography.
Catalog No. 50-240-8537