Learn More
Sino Biological Recombinant Human Factor X Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11076H08B500
Encompass_Preferred
A DNA sequence encoding the full length of human coagulation factor X (NP_000495.1) (Met 1-Lys 488) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4875