Learn More
Sino Biological Recombinant Anti-Factor XI Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10302R006H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Coagulation Factor XI/F11 (rh Coagulation Factor XI/F11; Catalog#10302-H08H; NP_000119.1; Met1-Val625), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-7881