Learn More
Sino Biological Recombinant Human JAM-A/F11 Receptor Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Catalog No. 502992890
Encompass_Preferred
A DNA sequence encoding the Human F11R (NP_058642.1) (Met1-Ala242) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2890 Supplier Sino Biological Supplier No. 10198H49HB100
A DNA sequence encoding the Human F11R (NP_058642.1) (Met1-Ala242) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.