Learn More
Sino Biological Anti-JAM-A/F11 Receptor Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10198MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Junctional Adhesion Molecule A (rh Junctional Adhesion Molecule A; Catalog#10198-H08H; NP_058642.1; Met1-Ala242). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1375